



Cyclohexane : Ethyl acetate mixture (50:50) (v/v) for pesticide residue analysis
Catalog # VWRS83957.320
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (Cyclohexane) (V/V):49.5 50.5 %
- Assay (Ethyl acetate) (V/V):49.5 50.5 %
- Lindane (GC/ECD) max. 5 ng/l:Passes test
Specification Test Results
About this item
:
UN:1993
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient