VWR® Maximum SF Cleanroom Lab Coats
VWR® Maximum SF lab coats are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness, and comfort properties.
- Tapered collar, inset sleeves, snap closure, elasticated cuffs and three pockets
- Superior fluid and particulate barrier
- Lowest level of particle shedding: Level I Helmke drum classification
- Light chemical splash resistance
- Improved breathability to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
Tested and manufactured in an ISO certified facility under stringent process controls. Lot controlled and validated through independent laboratory testing.
- Catalog No:
- 113-1138
- 113-1139
- 113-1140
- 113-1141
- 113-1143
- 113-1142
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
Frequently Bought Together