Tritylium hexafluorophosphate, Sigma-Aldrich®
Catalog # SIAL164569-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:388.29 g/mol
Melting Pt:166…167 °C
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:207-112-0
CAS Number:437-17-2
Formula:C₁₉H₁₅F₆P
MDL Number:MFCD00013121