Order Entry
Czech Republic
ContactUsLinkComponent
4,4'-Dimethoxybenzophenone, Sigma-Aldrich®
4,4'-Dimethoxybenzophenone, Sigma-Aldrich®
Catalog # SIAL141984-25G
Supplier:  Merck
CAS Number:  
4,4'-Dimethoxybenzophenone, Sigma-Aldrich®
Catalog # SIAL141984-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :242.27 g/mol
Boiling Pt:180…240 °C (13 mmHg)
Melting Pt:142…145 °C
Density:1.259 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:202-028-0
CAS Number:90-96-0
Formula:C₁₅H₁₄O₃
MDL Number:MFCD00008404