

Ethanolamine 50% in aqueous solution
Supplier: VWR Collection
Certificates
About this item
CAS Number:141-43-5
:
Formula:NH₂CH₂CH₂OH
MDL Number:MFCD00008183
Boiling Pt:∼170 °C (1013 hPa)
Melting Pt:1 °C
Density:1.01 g/cm³ (20 °C)
EINECS:205-483-3
UN:2491
ADR:8,III
:61.08 g/mol
:GHS05
:GHS07
Frequently Bought Together