Penicillinase (beta-Lactamase) Typ IV (from Enterobacter cloacae)
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:9001-74-5
- Source:Enterobacter cloacae
- Storage conditions:Refrigerator
- Enzyme name:Penicillinase (beta-Lactamase), Typ IV
Specifications
Frequently Bought Together