di-Sodium β-glycerophosphate pentahydrate, Sigma-Aldrich®
Catalog # SIAL50020-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:306.11 g/mol
EINECS:212-464-3
CAS Number:13408-09-8
:819-83-0
Formula:C₃H₇Na₂O₆P·5H₂O
MDL Number:MFCD00002135