β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate 95+%, reduced
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1949720-50-6
:606-68-8
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O
MDL Number:MFCD03791273
:727.43 g/mol
Melting Pt:140…142 °C
Storage Temperature:Refrigerator
EINECS:210-123-3
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Water:≤8.0 % (K.F.)
- HPLC:≥95.0 %
Specification Test Results
Frequently Bought Together