Amberlite® IRN-78, Ion exchange resin cationic type (OH-) nuclear grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:11128-95-3
:
MDL Number:MFCD00145822
Merck Index:14,382
Storage Temperature:Ambient
Related Information
Frequently Bought Together