ContactUsLinkComponent
Lugol's iodine solution, Sigma-Aldrich®
Lugol's iodine solution, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Lugol's iodine solution, Sigma-Aldrich®
Supplier:  Merck
Certificates
Specification Test Results
  • Suitability for microscopy:
    passes test
  • gram-positve microorganisms:
    blue-violet
  • gram-negative microorganisms:
    pink to red