Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Lugol's iodine solution
Lugol's iodine solution
Lugol's iodine solution
Lugol's iodine solution
Katalogové číslo SCARLU00101000
Supplier:
Scharlau
CAS Number:
Expand image
Lugol's iodine solution
Katalogové číslo SCARLU00101000
Supplier:
Scharlau
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
:
Density:
1.01 g/cm³ (20 °C)
Storage Temperature:
Ambient
Product Details