Lugol's iodine solution, Sigma-Aldrich®
Katalogové číslo SIALL6146-1L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Density:1.01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient