Azomethine H sodium salt, Supelco®
: Merck
:5941-07-1
:
:C₁₇H₁₂NNaO₈S₂
:MFCD00042050
:445.41 g/mol
:> 200 °C
:M-Clarity™ quality level = MQ100
:Ambient
:227-698-1
- Solubility:(20 °C) slightly soluble
- Molar Mass:445.39 g/mol
- Bulk Density:500 kg/m3
Frequently Bought Together