
4'-Chloro-2'-hydroxyacetophenone ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6921-66-0
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00238557
:170.6
Melting Pt:27…30 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together