Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5'-Bromo-2'-hydroxyacetophenone
5'-Bromo-2'-hydroxyacetophenone, Sigma-Aldrich®
5'-Bromo-2'-hydroxyacetophenone, Sigma-Aldrich®
SIAL383406-10G
:
Merck
:
5'-Bromo-2'-hydroxyacetophenone, Sigma-Aldrich®
SIAL383406-10G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
215.05
:
M-Clarity™ quality level = MQ100
:
1450-75-5
:
C₈H₇BrO₂
:
MFCD00191850