Magnetic hotplate stirrer, MR Hei-Standard
Magnetic hotplate stirrer with electronic temperature control and analogue control dials for speed and temperature. Ceramic coated silumin plate provides excellent heat conduction and distribution, and is extremely resistant to scratches and chemicals. An independent safety circuit with separate circuit switches off the heating if the nominal hotplate temperature is exceeded by 25 °C. Electronic current limiting to protect the motor. TRIAC guarantees long service life.
- Extra safety control circuit and hotplate cut-out by two independent temperature sensors
- Medium temperature range: Up to 300 °C
IP protection class according to DIN EN 60529: IP 32
: IP protection class according to DIN EN 60529: IP 32
Frequently Bought Together