Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Köffler bench

Köffler bench

Köffler Bench
Katalogové číslo 621-2105
Dodavatel:   WAGNER MUNZ
Köffler bench
Katalogové číslo 621-2105
Dodavatel:  WAGNER MUNZ
Köffler bench
Köffler bench

Köffler Bench

Přejít na:
Price
65 610,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents

Rapid melting point measurement between 50 and 260 °C.

  • Assymetrically heated metal block
  • Pointer with scale to easily read the temperature
  • One point calibration with standard
  • Integrated controller

Power: 150 W

Specifikace
  • Description:
    Köffler Bench
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.