ContactUsLinkComponent
Laboratory blenders, Masticator
Laboratory blenders, Masticator
Supplier:  IUL Instruments SA
Certificates
undefined
Laboratory blenders, Masticator
Supplier:  IUL Instruments SA
Specifications
  • Catalog No:
  • 710-0104
  • 710-0101
  • 710-0100
  • 710-0103
  • 710-0102
  • 710-0105
  • Model:
  • Classic/Basic
  • Silver
  • Classic/Basic
  • Classic/Basic
  • Classic/Basic
  • Silver
  • Motor speed:
  • 1500 min⁻¹
  • 500…1500 min⁻¹
  • 1500 min⁻¹
  • 1500 min⁻¹
  • 1500 min⁻¹
  • 500…1500 min⁻¹
  • Reduction:
  • 1:6
  • 1:6
  • 1:6
  • 1:6
  • 1:6
  • 1:6
  • Strokes:
  • 8,5/s
  • 5/6/7/8/s
  • 8,5/s
  • 8,5/s
  • 8,5/s
  • 5/6/7/8/s
  • Timer:
  • 15 - 90 seconds; digital setting, continuous mode
  • 5/30/60/90/120/150/180/210/240 s; keyboard digital setting, continuous mode
  • 15 - 90 seconds; digital setting, continuous mode
  • 15 - 90 seconds; digital setting, continuous mode
  • 15 - 90 seconds; digital setting, continuous mode
  • 5/30/60/90/120/150/180/210/240 s; keyboard digital setting, continuous mode
  • Start/stop:
  • door activated
  • door activated
  • door activated
  • door activated
  • door activated
  • door activated
  • Power:
  • 270 W
  • 270 W
  • 270 W
  • 270 W
  • 270 W
  • 270 W
  • Motor:
  • Induction
  • Induction
  • Induction
  • Induction
  • Induction
  • Induction
  • W×D×H:
  • 240×400×260 mm
  • 240×400×300 mm
  • 240×400×260 mm
  • 240×400×260 mm
  • 240×400×260 mm
  • 240×400×300 mm
  • Weight:
  • 17 kg
  • 20 kg
  • 17 kg
  • 17 kg
  • 17 kg
  • 20 kg