ContactUsLinkComponent
Disinfectant solutions, Bacillol AF
Disinfectant solutions, Bacillol AF
  148-0138
undefined
Disinfectant solutions, Bacillol AF
  148-0138
  • Description:
    Rapid Disinfectant Solution, Bacillol AF
  • Seq:
    07
  • Packed:
    1 litre bottle
  • Pk:
    1 L
Alcohol based disinfectant, aldehyde-free and fragrance-free, suitable for spray wipe of all surfaces resistant to alcohol in the agro-food and pharmaceutical industries, in hospital environments or in medical practices. Suitable for application where rapid disinfection is required that leaves no residue.
Contents:
Propan-1-ol 450 mg/g
Propan-2-ol 250 mg/g
Ethanol 47 mg/g
 : Use biocides safely. Always read the label and product information before use.
Avantor recommend that you read the package insert and the instructions prior to each use of this product.