ContactUsLinkComponent
Accessories for immersion thermostats, Optima series
Accessories for immersion thermostats, Optima series
Catalog # 461-1001
Supplier:  GRANT INSTRUMENTS
undefined
Accessories for immersion thermostats, Optima series
Catalog # 461-1001
Supplier:  GRANT INSTRUMENTS
Specifications
  • Description:
    Clamp. Alternative to bridge plate allows attachment to vertical sided tanks. Tank can have a maximum wall thickness of 35 mm (rectangular) or 30 mm (circular, up to 30 mm diameter) and a capacity of up to 50 litres. For T100, TC120, TX150 and TXF200 immersion thermostats
  • Pk:
    1