O tomto produktu
Eppendorf rotors for centrifuges 5804(R) and 5810(R) precision meets power. Customise your centrifugation with varied capacities and speeds. Accessorise with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
- Accessories (adaptors and buckets) further customise your centrifuge setup
- Aerosol-tight options available
Experience precision in high-performance centrifugation with Eppendorf Rotors designed for centrifuge 5804/5804 R. These rotors offer diverse capacities and speeds, providing flexibility for various sample processing needs. Accessorise with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-performance applications, the centrifuge 5804 series is further optimised with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your centrifugation experience.
Specifikace
- Catalog No:
- 521-2073
- 521-1957
- 522-2454
- 521-2078
- 521-2076
- 521-0055
- 521-0046
- 521-2081
- 521-2080
- 521-0051
- 521-0054
- 521-0050
- 521-0049
- 521-0044
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001