Order Entry
Czech Republic
ContactUsLinkComponent
Blocks for cooling/heating microtube dry blocks, ThermoStat plus
Blocks for cooling/heating microtube dry blocks, ThermoStat plus
Catalog # 460-0099
Supplier:  EPPENDORF
undefined
Blocks for cooling/heating microtube dry blocks, ThermoStat plus
Catalog # 460-0099
Supplier:  EPPENDORF

Specifications

  • Description:
    Interchangeable thermoblock for 4 Eppendorf DualChip slides, with clamping plate
  • Seq:
    006
  • Max. revolutions:
    1400 min-1
  • Pk:
    1
  • For:
    70 °C max

Specifications

About this item