Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Wipes for general applications, Polytex®

Wipes for general applications, Polytex®

Wipes for general applications, Polytex®
Certificates
Wipes for general applications, Polytex®
Wipes for general applications, Polytex®

4 Options Available Below

Product Details & Documents

Soft cloths for wet wiping of sensitive surfaces.

  • 100% PP, reusable
  • Solvent resistant
  • Anti-static and fluff-free for universal lab applications and food areas
  • High tear resistance and good absorption capacity
Product Family Options

Product Information

  • Colour
    L×W
    Packed
    Availability
    Price
  • 115-2701
    60 g/m²
    60 g/m²
    White
    420×420 mm
    Pack of 35
    Log in for availability
    Log in for availability
    1 * 35 KS
    1 * 35 KS - 142,00 Kč
    142,00 Kč
    1 * 35 KS
       142,00 Kč

  • 115-2702
    60 g/m²
    60 g/m²
    White
    420×420 mm
    Pack of 420
    Log in for availability
    Log in for availability
    1 * 420 KS
    1 * 420 KS - 1 710,00 Kč
    1 710,00 Kč
    1 * 420 KS
       1 710,00 Kč

  • 115-2703
    75 g/m²
    75 g/m²
    Blue
    420×420 mm
    Pack of 35
    Log in for availability
    Log in for availability
    1 * 35 KS
    1 * 35 KS - 153,00 Kč
    153,00 Kč
    1 * 35 KS
       153,00 Kč

  • 115-2704
    75 g/m²
    75 g/m²
    Blue
    420×420 mm
    Pack of 420
    Log in for availability
    Log in for availability
    1 * 420 KS
    1 * 420 KS - 1 840,00 Kč
    1 840,00 Kč
    1 * 420 KS
       1 840,00 Kč

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.