Cleanroom wipes, pre-saturated, TechniSat®
Supplier: Texwipe
Certificates
About this item
These pre-wetted wipes are made of a fabric blend of 55% non woven hydroentangled cellulose and 45% polyester.
- Convenient wipes offer portion control and reduced potential of spillage in and out of the cleanroom
- Excellent bi-directional strength and a very low extractable level
- Ideal for cleaning environmental cleanroom surfaces during and following the production flow
- Suitable for wiping of production equipment requiring low solvent extractables and low particle generation
This fabric combines the highly absorbent properties of a natural fibre with the cleanliness and strength of a synthetic fibre. It offers low particle counts, low extractable levels and solvent compatibility. TechniSat® wipes are pre-wetted with isopropyl alcohol/deionised water solutions filtered to 0,2 μm. The wipes are packed in resealable FlexPacks, canisters or slider bags.
Colour: White
Suitable for use in Class 100 - 100000, ISO 5 - 8 cleanrooms.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together