Diagnostic vials, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
These clear PETG vials with white HDPE closures are ideal for sampling, storage and shipment of reagents or buffer solutions.
- Non-pyrogenic and non-cytotoxic
- Sterile to 10⁻⁶ SAL
- 20-415 lined white high-density polyethylene closure attached to PETG vial
- Leakproof when used with Nalgene closures
- Narrow mouth
These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.
Inner neck diameter: 11 mm
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Packed in shrink-wrapped trays with closures attached.
Specifications
- Catalog No:
- 391-0076
- 391-0075
- 391-0074
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement