Screw caps with liners, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
These PP screw caps have multi-purpose liners, which are made of three-ply co-extruded foamed low density PE core between two solid layers of low density PE.
- For use with neutral pH to mild acid and alkali solutions
Frequently Bought Together