Caps, LABOCAP, with grip
Dodavatel: LUDI
Certifikáty
O tomto produktu
Aluminium.
- Made of anodised aluminium with inner stainless steel spring
- Springs clamp against the outer wall of the vessel
- Anodised and coloured
- Autoclavable
With grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
Frequently Bought Together