Centrifuge tubes, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PPCO, round bottom and rimless, autoclavable.
- Translucent, excellent chemical resistance
- Withstand forces up to 50000×g
- More suitable for low temperatures than PP
Ordering information: 525-2859 and 525-2860 are lipped.
Specifications
- Catalog No:
- 525-2853
- 525-2859
- 525-2860
- 525-2855
- 525-2858
- 525-2857
- 525-2854
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement