ContactUsLinkComponent
Beaker tongs, stainless steel
Beaker tongs, stainless steel
Catalog # 231-4145
Supplier:  RSG
undefined
Beaker tongs, stainless steel
Catalog # 231-4145
Supplier:  RSG
Specifications
  • Length:
    330 mm
  • Pk:
    1
  • Spanning area:
    60 - 150 mm
Specifications
About this item
Stainless steel ¹⁸/₁₀, anti-magnetic. For easy, safe handling of beakers and similar containers. The jaws are wrapped in ceramic fibres to provide thermal insulation and to protect the containers they hold.
  • Corrosion resistant
  • Excellent thermal resistance, autoclavable