Order Entry
Czech Republic
ContactUsLinkComponent
Cleanroom tapes, double-sided
Cleanroom tapes, double-sided
Catalog # UTAP1510CL200-P3DU
Supplier:  ULTRATAPE
undefined
Cleanroom tapes, double-sided
Catalog # UTAP1510CL200-P3DU
Supplier:  ULTRATAPE

Specifications

  • Width:
    50,8 mm
  • Description:
    Double sided cleanroom tape, permanent adhesive
  • Seq:
    04
  • Pk:
    1 Roll

Specifications

About this item

These cleanroom tapes are made of polyester and are ideal for signs, name-plate mounting, or for connecting plastic sheeting pieces in cleanroom wall construction.

  • Disposable red Mylar™ liner
  • Permanent adhesive (will leave residue when removed)

Length: 32,9 m

Colour: Transparent