ContactUsLinkComponent
Sealing tape with dispenser, Nunc™
Sealing tape with dispenser, Nunc™
Catalog # 391-0113
   Low Manufacturing Impact
undefined
Sealing tape with dispenser, Nunc™
Catalog # 391-0113
   Low Manufacturing Impact
Specifications
  • Description:
    Sealing tape with dispenser
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1.000
Specifications
About this item
Polyester tape with acrylic adhesive. Easy peel, easy seal general-purpose tape with a faster application time than normal tape in loose sheets.
  • Temperature stable from −20 to +120 °C
  • Non-pierceable by pipette
Colour: clear
External L×W: 121×78 mm (plus handle 9×78 mm)