ContactUsLinkComponent
Adhesive seal, SILVERseal™
Adhesive seal, SILVERseal™
Katalogové číslo GREI676090_1200
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Adhesive seal, SILVERseal™
Katalogové číslo GREI676090_1200
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Specifications
  • Description:
    SILVERseal™
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Pk:
    1.200
Specifications
About this item
SILVERseal™ is a heat resistant, cold-resistant and pierceable aluminium foil, which is suitable for both PCR applications and sample storage.
  • Temperature resistant from −80 to 110 °C
  • Coated with an acrylate adhesive, which remains flat when removed from its paper backing, and can be easily applied to all microplate types
  • A double perforation makes it possible to tear off the projecting adhesive sealer and thus improves the stacking of sealed microplates
VIEWseal™ is an adhesive sealing film coated with a silicone adhesive, which only sticks when the film surface is pressed on. The film is easy to work with, even when wearing gloves (the film does not adhere to gloves).
AMPLIseal™ is a self-adhesive sealing film notable for its low autofluorescence, in particular in the wavelength range critical for Real Time PCR. It consists of a thick PP film coated with a transparent acrylate adhesive. The adhesive layer is protected by a peelable polyester film. The strongly adhering acrylate adhesive provides reliable sealing, thereby minimising evaporation but without influencing the PCR reaction or the fluorescence measurement.