ContactUsLinkComponent
Sealing film for PCR plates, Axygen® UltraClear
Sealing film for PCR plates, Axygen® UltraClear
Catalog # 732-7509
Supplier:  Corning
undefined
Sealing film for PCR plates, Axygen® UltraClear
Catalog # 732-7509
Supplier:  Corning
Specifications
  • Description:
    Sealing film, UltraClear
  • Pk:
    100
Specifications
About this item
For sealing 96 and 384-well PCR plates.
  • Film clarity allows for optical analysis during Real-Time PCR
  • Unique pressure sensitive adhesive that leaves no tacky residue
  • Functional temperature range −40 to +104 °C