Sealing film, Axygen® AxySeal
Supplier: Corning
Certificates
About this item
Polyester based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in ELISA assays. Suitable for sealing tissue culture plates, for short-term storage and incubation, for transport and the containment of biohazardous solutions.
- No edge effect in ELISA assays
- Functional temperature range of −40 to +104 °C
Frequently Bought Together