Orders LinkContactUsLinkComponent
Dissecting scissors
Dissecting scissors
Supplier:   HAMMACHER KARL
Dissecting scissors
Supplier:  HAMMACHER KARL
Certificates
Dissecting scissors
Dissecting scissors
3 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • Ideal for a wide range of laboratory applications
  • Acid-resistant
  • Autoclavable
  • Sharp-sharp
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Straight, sharp/sharp
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 560,00 Kč
    2 560,00 Kč
    1 * 1 KS
       2 560,00 Kč
  • Slightly curved, sharp/sharp
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 650,00 Kč
    2 650,00 Kč
    1 * 1 KS
       2 650,00 Kč
  • Curved, sharp/sharp
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 200,00 Kč
    3 200,00 Kč
    1 * 1 KS
       3 200,00 Kč