Order Entry
Czech Republic
ContactUsLinkComponent
Racks for scintillation vials, WHEATON®
Racks for scintillation vials, WHEATON®
  212-6190
undefined
Racks for scintillation vials, WHEATON®
  212-6190

 

  • Hole Ø:
    28 mm
  • Description:
    Rack for scintillation vials
  • Pk:
    5

 

 

PP.

Rack for 5×10 scintillation vials with diameter 28 mm.
W×D×H: 320×170×35 mm