Pipette tips, universal, Axygen®
Supplier: Corning
Certificates
About this item
These universal pipette tips are compatible with most micropipettes.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
Ordering information: Available loose or in racks, sterile or non sterile versions.
Specifications
- Catalog No:
- 613-1271
- 613-1270
- 613-0965
- 613-1269
- 613-3830
- 615-0029
- 613-3829
- 613-3828
- 613-3827
- 613-1297
- 613-1267
- 613-0659
- 613-0658
- 613-1293
- 613-1263
- 613-1292
- 613-0657
- 613-3826
- 613-1291
- 613-1290
- 613-1289
- AXYGTR-222-C-STK-S
- 613-1343
- 613-1340
- 613-1341
- 613-1342
- 613-0656
- 613-0901
- 613-1288
- 613-0648
- 613-0651
- 613-0647
- 613-0629
- 613-0650
- 613-0646
- 613-0649
- 613-1336
- 613-1338
- 613-0642
- 613-1320
- 613-0628
- 613-1334
- 613-0663
- 613-0668
- 613-0667
- 613-1277
- 613-1276
- 613-0645
- 613-0666
- 613-1309
- 613-0644
- 613-1272
- 613-0630
- 613-0641
- 613-1299
- 613-0670
- 613-0643
- 613-0669
- 613-0640
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa