ContactUsLinkComponent
Pipette tray for drawer
Pipette tray for drawer
Catalog # 612-1805
Supplier:  KARTELL
undefined
Pipette tray for drawer
Catalog # 612-1805
Supplier:  KARTELL
Specifications
  • Description:
    Pipette tray for drawer
  • Pk:
    1
Specifications
About this item
PVC, white.
  • Designed to hold about 30 pipettes
  • Four compartments to accommodate 1, 2, 5, 10 ml pipettes
W×D×H: 425×300×30 mm.