ContactUsLinkComponent
VWR®, Safety Bulb Pipette Fillers
VWR®, Safety Bulb Pipette Fillers
Catalog # 612-2703
Supplier:  VWR Collection
undefined
VWR®, Safety Bulb Pipette Fillers
Catalog # 612-2703
Supplier:  VWR Collection
Specifications
  • Colour:
    Red/white
  • Description:
    Safety bulb pipette fillers, 15 ml
  • Seq:
    01
  • Pk:
    3
Specifications
About this item
Red rubber with white plastic connector and silicone conical adapter, autoclavable.
  • For large and small capacity pipettes
  • Conical adapter connects pipettes more securely, avoids breakages because it requires less force to fit on the bulb
  • Elastic cord for attaching the bulb to a reagent vessel or to a user's wrist
Capacity: 15 ml