Order Entry
Czech Republic
ContactUsLinkComponent
Pipette controller, Bibbyjet Pro
Pipette controller, Bibbyjet Pro
Catalog # 613-4180
Supplier:  Cole Parmer
undefined
Pipette controller, Bibbyjet Pro
Catalog # 613-4180
Supplier:  Cole Parmer

Specifications

  • Description:
    Bibbyjet Pro with charger for UK and Ireland
  • Seq:
    02
  • Pk:
    1

Specifications

About this item

Pipette controller for glass or plastic pipettes from 0,1 to 100 ml.

  • Just two buttons for every function
  • One full working day of pipetting without recharging
  • Built-in non return valve combined with a membrane filter for efficient protection against penetration of liquids
  • Specially designed silicone adapter allowing a tight fit for all glass or plastic pipettes from 0,1 to 100 ml

Powerful, lightweight and handy - the automatic Bibbyjet Pro has just two function buttons to give you complete control over filling and delivery. Gently or powerfully, the built-in dual controls allow you to take up liquid slowly or quickly, whatever your needs.

Ordering information: Supplied with wall support and two spare 0,2 ml membrane filters.