Graduated pipettes, SILBERBRAND ETERNA, type 3, Class B, total delivery
: Brand
AR-Glas®.
- Calibrated to deliver (TD, Ex)
- Marks and inscriptions in ETERNA amber stain
- Marked pipettes (*) suitable for cotton plugs
DIN EN ISO 835
Length: 360 ±10 mm except for 25 ml pipettes: 450 ±10 mm.
: DIN EN ISO 835.
Frequently Bought Together