ContactUsLinkComponent
Mops, Anticon® SuperSorb
Mops, Anticon® SuperSorb
Catalog # 129-0216
Supplier:  CONTEC
undefined
Mops, Anticon® SuperSorb
Catalog # 129-0216
Supplier:  CONTEC
Specifications
  • Description:
    Anticon® SuperSorb mop
  • Pk:
    1
Specifications
About this item
Made from 100% continuous filament polyester with two-yarn system for superior sorbency. Designed for use with commercially available handles and bucket-wringer systems.
  • For wet mopping in critical areas where maximum sorbency is required for applying cleaning and disinfectant solutions
  • Mop is durable, can be laundered for repeated use
  • Autoclavable for sterile applications
Packaging: Individually packed.