Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, PCR racks, reversible
VWR®, PCR racks, reversible
Catalog # 211-0198
Supplier:  VWR Collection
undefined
VWR®, PCR racks, reversible
Catalog # 211-0198
Supplier:  VWR Collection

Specifications

  • Colour:
    Assorted colours
  • Description:
    PCR racks
  • Seq:
    01
  • Pk:
    5

Specifications

About this item

PP.

  • PCR side of the rack has 168 wells that can hold 8-well or 12-well tube strips, or one 0,2 ml tube per well
  • Opposite side of the rack has 40 wells that can hold 0,5 ml tubes
  • Both sides of the rack have 12 wells that can hold 1,5 ml tubes
  • Simple to use, removable hinged lid snaps in place on either side of the rack

Designed to hold 0,2; 0,5 or 1,5 ml tubes.

Ordering information: Assorted pack includes one each of blue, green, purple, yellow and orange.