ContactUsLinkComponent
Storage plate cap strips
Storage plate cap strips
Catalog # 732-4922
   Low Manufacturing Impact
undefined
Storage plate cap strips
Catalog # 732-4922
   Low Manufacturing Impact
Specifications
  • Description:
    Storage plate 8-cap strips
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    120
Specifications
About this item
Strips of eight caps for sealing storage plates and racked cluster tubes. The strips are compatible with Thermo Scientific microplates, 0,8 ml storage plates, 1,2 ml round well storage plates and 1,2 ml cluster tubes ranges.
  • Each cap is individually numbered
  • Simultaneous sealing of 8 wells
  • End tabs for ease of removal
  • Low profile for increased well volume