Storage plates, 96-well, V-bottomed
Catalog # 732-4942
Supplier: THERMO MATRIX TECHNOLOGIES
Specifications
- Description:V-bottomed 96-well storage plate
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Pk:100
Specifications
About this item
PP.
- V-bottomed well design for optimal sample recovery
- Reinforced skirt for robotic manipulation
- 0,2 ml maximum well volume