Storage plates, 1536-well, MASTERBLOCK®
Supplier: Greiner Bio-One
Certificates
About this item
PP, without lid. Ideal for high throughput screening applications.
- Biologically inert, resistant to commonly used solvents (such as DMSO, acetonitrile or acetone) and temperature resistant (−196 °C to +121 °C)
- Uniform external dimensions and tolerances
- Can be sealed with adhesive films and heat seals
- Alphanumeric well coding
- Working volume: 3 to 15 µl
Colour: Natural
Specifications
- Catalog No:
- 737-3532
- 738-0081
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Specifications
Frequently Bought Together