Sterilin® Microtitre Plates, 96 Well
: Sterilin
PS, non treated. Used for serology and microbiology. Compatible with automated plate readers and washers.
- Suitable for use at a wavelength of 340 nm
- Condensation rings on the lid (supplied separately) together with raised well rims on the base help minimise the risk of contamination from surrounding wells
- Frosted write-on area on end wall, orientation corners and alpha numeric labelling ensure easy sample identification
- Hanging well design ensures even temperature distribution around each well
- Manufactured from virgin polystyrene for excellent optical clarity
- Aseptically manufactured without surface treatment for a medium binding surface
- Irradiated for sterility and a more hydrophilic surface, providing high binding
: CE marked in accordance with the European Directive 98/72/EC.
Frequently Bought Together