About this item
These ambidextrous gauntlets are manufactured from natural rubber that provides good mechanical properties such as tensile strength, perforation and abrasion resistance.
- Good flexibility and low modulus
- Low static charge
- Very high resistance to cold conditions (down to −40 °C)
- Combination of flexibility and mechanical strength offers protection together with high dexterity for the user
0,4 mm thickness: EN 388 (X100), EN 374-1, EN 421; Cat. III
0,6 mm thickness: EN 388 (X111), EN 374-1, EN 421; Cat. III
0,6 mm thickness: EN 388:2016 (XX11X), EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 421; Cat. III