Glove liners
Supplier: WELLS LAMONT
Certificates
About this item
Ambidextrous liners are constructed of high-stretch nylon for maximum comfort and superior tactile sensitivity. They provide a physical barrier to protect hands from the irritating effects of over exposure to latex, vinyl and nitrile thin film gloves.
- Can also be used as gowning or inspection gloves
- Available in full-finger and half-finger styles
- Launderable
Frequently Bought Together