ContactUsLinkComponent
Latex gloves, Semperclean MC
Latex gloves, Semperclean MC
Dodavatel:  SEMPERIT TECHN PROD
Certifikáty
undefined
Latex gloves, Semperclean MC
Dodavatel:  SEMPERIT TECHN PROD
Specifikace
  • Catalog No:
  • 112-5700
  • 112-3131
  • 112-5701
  • 112-3130
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • EN norm:
  • EN 455, EN 374
  • EN 455, EN 374
  • EN 455, EN 374
  • EN 455, EN 374
Specifikace
Související informace
Frequently Bought Together